Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGTGGATGCGGACCCGGATGGC[A/G]TGCTGGCGGATGAAGGCAATCTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604992 | ||||||||||||||||||||
Literature Links: |
SRL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRL - sarcalumenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098814.1 | 1067 | Silent Mutation | NP_001092284.1 | |||
NM_001323667.1 | 1067 | Silent Mutation | NP_001310596.1 | |||
NM_001323668.1 | 1067 | Silent Mutation | NP_001310597.1 | |||
XM_017023527.1 | 1067 | Silent Mutation | XP_016879016.1 | |||
XM_017023528.1 | 1067 | Silent Mutation | XP_016879017.1 | |||
XM_017023529.1 | 1067 | Silent Mutation | XP_016879018.1 |