Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAAGCCCACTCCTCCGCAGGT[A/G]GACGAGGAGGTGCTCAAGTGCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608300 MIM: 600781 | ||||||||||||||||||||
Literature Links: |
NAGS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAGS - N-acetylglutamate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153006.2 | 319 | Silent Mutation | GTA,GTG | V,V 143 | NP_694551.1 | |
XM_011524438.1 | 319 | Silent Mutation | GTA,GTG | V,V 143 | XP_011522740.1 | |
XM_011524439.2 | 319 | UTR 5 | XP_011522741.1 |
PYY - peptide YY | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004160.5 | 319 | Intron | NP_004151.3 | |||
XM_011525035.1 | 319 | Intron | XP_011523337.1 |
TMEM101 - transmembrane protein 101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |