Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAGCACGGGCAGCCGGGCCAG[A/G]TTAAGCACCCCCAGGCTCATCATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103270 | ||||||||||||||||||||
Literature Links: |
FADS6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FADS6 - fatty acid desaturase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178128.5 | 846 | Silent Mutation | AAC,AAT | N,N 285 | NP_835229.3 | |
XM_005257224.4 | 846 | Silent Mutation | AAC,AAT | N,N 285 | XP_005257281.2 | |
XM_017024458.1 | 846 | Silent Mutation | AAC,AAT | N,N 267 | XP_016879947.1 |
FDXR - ferredoxin reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |