Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCAACAAGGCCTTCCTGGACTCC[C/G]GAACCAGGTACGTGAGACACACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 150205 | ||||||||||||||||||||
Literature Links: |
LPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPO - lactoperoxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160102.1 | 349 | Intron | NP_001153574.1 | |||
NM_006151.2 | 349 | Silent Mutation | CGA,GGA | R,G 53 | NP_006142.1 | |
XM_011524808.2 | 349 | Missense Mutation | CGA,GGA | R,G 53 | XP_011523110.1 | |
XM_011524810.2 | 349 | Intron | XP_011523112.1 |