Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTGCAGCTCTTCCGGAGTCAC[C/G]TGCAGCCCCTTTTGGCTGAGGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603730 | ||||||||||||||||||||
Literature Links: |
SPHK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPHK1 - sphingosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142601.1 | 504 | Missense Mutation | CTG,GTG | L,V 38 | NP_001136073.1 | |
NM_001142602.1 | 504 | Missense Mutation | CTG,GTG | L,V 38 | NP_001136074.1 | |
NM_021972.3 | 504 | Missense Mutation | CTG,GTG | L,V 52 | NP_068807.2 | |
NM_182965.2 | 504 | Missense Mutation | CTG,GTG | L,V 124 | NP_892010.2 | |
XM_005257766.2 | 504 | Missense Mutation | CTG,GTG | L,V 38 | XP_005257823.1 |
UBE2O - ubiquitin conjugating enzyme E2 O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |