Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCTCTTCTCTCAGGTCACTCTA[C/T]ATGACATGATCCTGAAAAATATCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602268 MIM: 603735 MIM: 605129 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AOC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AOC2 - amine oxidase, copper containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AOC3 - amine oxidase, copper containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME3 - proteasome activator subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267045.1 | 1184 | Missense Mutation | CAT,TAT | H,Y 243 | NP_001253974.1 | |
NM_005789.3 | 1184 | Missense Mutation | CAT,TAT | H,Y 232 | NP_005780.2 | |
NM_176863.2 | 1184 | Missense Mutation | CAT,TAT | H,Y 245 | NP_789839.1 | |
XM_011524177.2 | 1184 | Missense Mutation | CAT,TAT | H,Y 171 | XP_011522479.1 |