Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAACTACTCACAGAGCCAGGAA[A/G]AAATGAATGCGTTGGTATTGCCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDYE4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPDYE4 - speedy/RINGO cell cycle regulator family member E4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128076.1 | 1520 | Silent Mutation | TTC,TTT | F,F 158 | NP_001121548.1 | |
XM_005256632.4 | 1520 | Silent Mutation | TTC,TTT | F,F 158 | XP_005256689.1 | |
XM_005256633.4 | 1520 | Silent Mutation | TTC,TTT | F,F 158 | XP_005256690.1 |