Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGGATGGTGGCACAGGCACTG[A/C]ACAGGCTGGGGCCTTTTGACGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603527 MIM: 610016 MIM: 613487 MIM: 607896 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPH1 - diphthamide biosynthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001383.3 | 1503 | UTR 3 | NP_001374.3 |
MIR132 - microRNA 132 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR212 - microRNA 212 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OVCA2 - ovarian tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080822.2 | 1503 | Missense Mutation | AAC,CAC | N,H 107 | NP_543012.1 |