Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCTGCAATCATGACCTCGATCT[C/T]CCCCAAGGTGAGGTCTTCTAGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAT9 - N-acetyltransferase 9 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305077.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | NP_001292006.1 | |
NM_001305078.1 | 407 | Missense Mutation | AAG,GAG | K,E 105 | NP_001292007.1 | |
NM_001305079.1 | 407 | Missense Mutation | AAG,GAG | K,E 105 | NP_001292008.1 | |
NM_001305080.1 | 407 | Missense Mutation | AAG,GAG | K,E 105 | NP_001292009.1 | |
NM_001305081.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | NP_001292010.1 | |
NM_001305082.1 | 407 | Missense Mutation | AAG,GAG | K,E 110 | NP_001292011.1 | |
NM_001305083.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | NP_001292012.1 | |
NM_001305084.1 | 407 | Missense Mutation | AAG,GAG | K,E 105 | NP_001292013.1 | |
NM_001305085.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | NP_001292014.1 | |
NM_001305086.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | NP_001292015.1 | |
NM_001305087.1 | 407 | Missense Mutation | AAG,GAG | K,E 80 | NP_001292016.1 | |
NM_001305088.1 | 407 | Missense Mutation | AAG,GAG | K,E 74 | NP_001292017.1 | |
NM_015654.4 | 407 | Missense Mutation | AAG,GAG | K,E 105 | NP_056469.2 | |
XM_005257212.4 | 407 | Missense Mutation | AAG,GAG | K,E 74 | XP_005257269.1 | |
XM_006721813.3 | 407 | Missense Mutation | AAG,GAG | K,E 74 | XP_006721876.1 | |
XM_011524607.2 | 407 | Missense Mutation | AAG,GAG | K,E 104 | XP_011522909.1 | |
XM_017024437.1 | 407 | Missense Mutation | AAG,GAG | K,E 80 | XP_016879926.1 | |
XM_017024438.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | XP_016879927.1 | |
XM_017024439.1 | 407 | Missense Mutation | AAG,GAG | K,E 104 | XP_016879928.1 | |
XM_017024440.1 | 407 | Missense Mutation | AAG,GAG | K,E 74 | XP_016879929.1 | |
XM_017024441.1 | 407 | Missense Mutation | AAG,GAG | K,E 74 | XP_016879930.1 |
SLC9A3R1 - SLC9A3 regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM104 - transmembrane protein 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |