Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGAAGGGACCATGTGGCTGCCT[C/T]GGGCTCTGTTGCTTCTCTGGGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD300A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD300A - CD300a molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256841.1 | 357 | Missense Mutation | CGG,TGG | R,W 5 | NP_001243770.1 | |
NM_007261.3 | 357 | Missense Mutation | CGG,TGG | R,W 5 | NP_009192.2 | |
XM_005256990.2 | 357 | Missense Mutation | CGG,TGG | R,W 5 | XP_005257047.1 | |
XM_005256991.4 | 357 | Missense Mutation | CGG,TGG | R,W 5 | XP_005257048.1 | |
XM_005256992.4 | 357 | Intron | XP_005257049.1 | |||
XM_006721656.3 | 357 | Intron | XP_006721719.1 | |||
XM_011524254.2 | 357 | Missense Mutation | CGG,TGG | R,W 5 | XP_011522556.1 |