Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGTGGAGCAGTTCTGCATGGC[A/G]GAAGGGCCCAGGGGCACCCGGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF18B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF18B - kinesin family member 18B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001264573.1 | 2281 | Silent Mutation | TCC,TCT | S,S 716 | NP_001251503.1 | |
NM_001265577.1 | 2281 | Silent Mutation | TCC,TCT | S,S 704 | NP_001252506.1 | |
XM_011524385.2 | 2281 | Silent Mutation | TCC,TCT | S,S 725 | XP_011522687.1 | |
XM_011524386.2 | 2281 | Silent Mutation | TCC,TCT | S,S 716 | XP_011522688.1 | |
XM_011524387.2 | 2281 | Silent Mutation | TCC,TCT | S,S 716 | XP_011522689.1 | |
XM_011524388.2 | 2281 | Silent Mutation | TCC,TCT | S,S 713 | XP_011522690.1 | |
XM_011524389.2 | 2281 | Silent Mutation | TCC,TCT | S,S 725 | XP_011522691.1 | |
XM_011524390.2 | 2281 | Silent Mutation | TCC,TCT | S,S 690 | XP_011522692.1 | |
XM_011524391.2 | 2281 | Intron | XP_011522693.1 |
MIR6783 - microRNA 6783 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |