Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCGGCAAGAGTAGGAAGTGACA[A/C]AGGCTGGGCGGCGCAGGATGGAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KRTAP16-1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTAP16-1 - keratin associated protein 16-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146182.1 | 1313 | Missense Mutation | TGT,TTT | C,F 438 | NP_001139654.1 |
KRTAP17-1 - keratin associated protein 17-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP29-1 - keratin associated protein 29-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |