Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCGAGTTCGGGAAGCAGTGCA[A/C]TTTCTCGGACCGCCAGGCCGAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605483 | ||||||||||||||||||||
Literature Links: |
DNAI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAI2 - dynein axonemal intermediate chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172810.1 | 183 | Missense Mutation | AAT,ACT | N,T 19 | NP_001166281.1 | |
NM_023036.4 | 183 | Missense Mutation | AAT,ACT | N,T 19 | NP_075462.3 | |
XM_011525125.2 | 183 | Missense Mutation | AAT,ACT | N,T 19 | XP_011523427.1 |