Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCGCCGCCACTGCCGCCGCCGC[C/T]GCCGCCGCCTGGGACCTGCTTCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935 | ||||||||||||||||||||
Literature Links: |
EXOC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC7 - exocyst complex component 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALR2 - galanin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003857.3 | 69 | Intron | NP_003848.1 | |||
XM_011525427.2 | 69 | Intron | XP_011523729.1 |
SRP68 - signal recognition particle 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260502.1 | 69 | Missense Mutation | AGC,GGC | S,G 12 | NP_001247431.1 | |
NM_001260503.1 | 69 | Intron | NP_001247432.1 | |||
NM_014230.3 | 69 | Missense Mutation | AGC,GGC | S,G 12 | NP_055045.2 |
ZACN - zinc activated ion channel | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |