Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGTTGGTTTCTGCTTCCTCAC[C/T]AACCAGTGGGCAGTCACCAACCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603926 MIM: 188300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf99 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf99 - chromosome 17 open reading frame 99 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGR2 - synaptogyrin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320523.1 | 453 | Intron | NP_001307452.1 | |||
NM_004710.4 | 453 | Silent Mutation | ACC,ACT | T,T 127 | NP_004701.1 | |
XM_005257792.3 | 453 | Intron | XP_005257849.1 |
TK1 - thymidine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |