Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTATTTGCCCTTTTCCTTTCC[A/T]ACTTGGCGAGATTTGGCTTTCCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603704 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRBA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRBA2 - KRAB-A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304947.1 | 480 | Intron | NP_001291876.1 | |||
NM_213597.3 | 480 | Intron | NP_998762.1 | |||
XM_005256443.2 | 480 | Intron | XP_005256500.1 | |||
XM_011523643.1 | 480 | Intron | XP_011521945.1 | |||
XM_011523644.1 | 480 | Intron | XP_011521946.1 | |||
XM_017024163.1 | 480 | Intron | XP_016879652.1 | |||
XM_017024164.1 | 480 | Intron | XP_016879653.1 | |||
XM_017024165.1 | 480 | Intron | XP_016879654.1 |
RPL26 - ribosomal protein L26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000987.3 | 480 | Silent Mutation | NP_000978.1 | |||
NM_001315530.1 | 480 | Intron | NP_001302459.1 | |||
NM_001315531.1 | 480 | Intron | NP_001302460.1 |