Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTCCAGTCTGGGTGTCCCAGC[A/G]AGCCCCTGCAGGGACAGTATGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610063 MIM: 614942 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH17 - dynein axonemal heavy chain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173628.3 | 6098 | Missense Mutation | NP_775899.3 | |||
XM_011525416.2 | 6098 | Missense Mutation | XP_011523718.1 | |||
XM_017025261.1 | 6098 | Missense Mutation | XP_016880750.1 |
PGS1 - phosphatidylglycerophosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024419.4 | 6098 | UTR 3 | NP_077733.3 | |||
XM_011525487.2 | 6098 | UTR 3 | XP_011523789.1 | |||
XM_011525488.2 | 6098 | UTR 3 | XP_011523790.1 | |||
XM_011525489.2 | 6098 | UTR 3 | XP_011523791.1 | |||
XM_017025357.1 | 6098 | UTR 3 | XP_016880846.1 | |||
XM_017025358.1 | 6098 | UTR 3 | XP_016880847.1 | |||
XM_017025359.1 | 6098 | UTR 3 | XP_016880848.1 | |||
XM_017025360.1 | 6098 | UTR 3 | XP_016880849.1 | |||
XM_017025361.1 | 6098 | UTR 3 | XP_016880850.1 | |||
XM_017025362.1 | 6098 | UTR 3 | XP_016880851.1 | |||
XM_017025363.1 | 6098 | UTR 3 | XP_016880852.1 | |||
XM_017025364.1 | 6098 | UTR 3 | XP_016880853.1 | |||
XM_017025365.1 | 6098 | UTR 3 | XP_016880854.1 | |||
XM_017025366.1 | 6098 | UTR 3 | XP_016880855.1 |