Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607435 MIM: 612070 MIM: 612071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERAL1 - Era like 12S mitochondrial rRNA chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317985.1 | 490 | Missense Mutation | ATG,GTG | M,V 138 | NP_001304914.1 | |
NM_001317986.1 | 490 | Missense Mutation | ATG,GTG | M,V 138 | NP_001304915.1 | |
NM_005702.3 | 490 | Missense Mutation | ATG,GTG | M,V 138 | NP_005693.1 |
FAM222B - family with sequence similarity 222 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR144 - microRNA 144 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR451A - microRNA 451a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR451B - microRNA 451b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4732 - microRNA 4732 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |