Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGTCAAAACTGTCCGGGGCCAA[C/G]TTTTCGAGGGACTTCATTCTTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMCR8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMCR8 - Smith-Magenis syndrome chromosome region, candidate 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144775.2 | 666 | Missense Mutation | AAC,AAG | N,K 52 | NP_658988.2 |
TOP3A - topoisomerase (DNA) III alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320759.1 | 666 | Intron | NP_001307688.1 | |||
NM_004618.4 | 666 | Intron | NP_004609.1 | |||
XM_011524001.2 | 666 | Intron | XP_011522303.1 |