Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATTTCTTCTCTGGGTTGGCACA[C/T]ACTTGGCGGTTCTTTCGGGTGACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 187011 | ||||||||||||||||||||
Literature Links: |
CCL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL5 - C-C motif chemokine ligand 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278736.1 | 284 | Missense Mutation | ATG,GTG | M,V 100 | NP_001265665.1 | |
NM_002985.2 | 284 | Silent Mutation | GTA,GTG | V,V 72 | NP_002976.2 |
HEATR9 - HEAT repeat containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |