Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGATCCATTCCTTCTCGGTCA[A/G]CTGCTTAACTCTGTTATCTTTCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614956 | ||||||||||||||||||||
Literature Links: |
SLFN12L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLFN12L - schlafen family member 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195790.1 | 7799 | Silent Mutation | CTG,TTG | L,L 350 | NP_001182719.1 | |
XM_017024005.1 | 7799 | Silent Mutation | CTG,TTG | L,L 379 | XP_016879494.1 | |
XM_017024006.1 | 7799 | Silent Mutation | CTG,TTG | L,L 350 | XP_016879495.1 | |
XM_017024007.1 | 7799 | Silent Mutation | CTG,TTG | L,L 350 | XP_016879496.1 | |
XM_017024008.1 | 7799 | Silent Mutation | CTG,TTG | L,L 332 | XP_016879497.1 | |
XM_017024009.1 | 7799 | Silent Mutation | CTG,TTG | L,L 332 | XP_016879498.1 |