Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGCTGGCAACAGCGCAGCAAG[G/T]ATCGGTAGAGCTGCAGTGGCCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LYRM9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYRM9 - LYR motif containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076680.1 | 141 | Missense Mutation | TAC,TCC | Y,S 19 | NP_001070148.1 | |
XM_011524491.2 | 141 | Missense Mutation | TAC,TCC | Y,S 45 | XP_011522793.1 | |
XM_017024325.1 | 141 | Missense Mutation | TAC,TCC | Y,S 19 | XP_016879814.1 |