Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGTGTTGAAAATTCCAACAGCT[C/T]TATCCTTGTCTGAAAGCTTGGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR3A1 - olfactory receptor family 3 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002550.2 | 824 | Missense Mutation | AAA,AGA | K,R 275 | NP_002541.2 | |
XM_017024695.1 | 824 | Intron | XP_016880184.1 | |||
XM_017024696.1 | 824 | Intron | XP_016880185.1 |
OR3A2 - olfactory receptor family 3 subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002551.3 | 824 | Intron | NP_002542.3 | |||
XM_017024697.1 | 824 | Intron | XP_016880186.1 | |||
XM_017024698.1 | 824 | Intron | XP_016880187.1 |