Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATGGGGCCCGGAGGATGGAAC[A/G]GTGGGGCCCTGAAGGGCGATGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PELP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PELP1 - proline, glutamate and leucine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278241.1 | 2217 | Missense Mutation | CCG,CTG | P,L 517 | NP_001265170.1 | |
NM_014389.2 | 2217 | Missense Mutation | CCG,CTG | P,L 714 | NP_055204.3 |