Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCTGCATGTCGTGGTCATTGG[G/T]CTGGACTCTGCTGGAAAGACCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL4D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL4D - ADP ribosylation factor like GTPase 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001661.3 | 193 | Silent Mutation | GGG,GGT | G,G 28 | NP_001652.2 | |
XM_011524782.2 | 193 | Silent Mutation | GGG,GGT | G,G 28 | XP_011523084.1 |