Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGCTTAGATTCCAACATGCCA[C/T]GGGAGGAGTACCTGCCTGGGCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612816 | ||||||||||||||||||||
Literature Links: |
MBTD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBTD1 - mbt domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP18 - UTP18, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016001.2 | 597 | Intron | NP_057085.2 | |||
XM_006721930.3 | 597 | Missense Mutation | ACG,ATG | T,M 190 | XP_006721993.2 | |
XM_011524870.2 | 597 | Missense Mutation | ACG,ATG | T,M 190 | XP_011523172.1 |