Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCAAACCCAAAGCCAGGTTGAC[A/G]CCCAAAATACAGAAACTTCTTAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610347 | ||||||||||||||||||||
Literature Links: |
C18orf21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C18orf21 - chromosome 18 open reading frame 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201474.1 | 381 | UTR 5 | NP_001188403.1 | |||
NM_001201475.1 | 381 | UTR 5 | NP_001188404.1 | |||
NM_001201476.1 | 381 | Silent Mutation | ACA,ACG | T,T 66 | NP_001188405.1 | |
NM_031446.4 | 381 | Silent Mutation | ACA,ACG | T,T 66 | NP_113634.3 | |
XM_005258364.4 | 381 | Silent Mutation | ACA,ACG | T,T 72 | XP_005258421.1 |
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |