Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTAATTCATGAGGCAACTCATG[A/G]CAGAACACAGCAACAGAAGTACTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608731 | ||||||||||||||||||||
Literature Links: |
SLC39A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC39A6 - solute carrier family 39 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099406.1 | 2625 | Silent Mutation | TGC,TGT | C,C 359 | NP_001092876.1 | |
NM_012319.3 | 2625 | Silent Mutation | TGC,TGT | C,C 634 | NP_036451.3 | |
XM_011525900.1 | 2625 | Silent Mutation | TGC,TGT | C,C 634 | XP_011524202.1 | |
XM_011525901.1 | 2625 | Silent Mutation | TGC,TGT | C,C 634 | XP_011524203.1 |