Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATTCAGATTCATGTTTGCTTA[A/G]AGACCCAATGAGGCAGAACCTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607973 | ||||||||||||||||||||
Literature Links: |
NETO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NETO1 - neuropilin and tolloid like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201465.1 | 1867 | Silent Mutation | CTA,TTA | L,L 521 | NP_001188394.1 | |
NM_138966.3 | 1867 | Silent Mutation | CTA,TTA | L,L 521 | NP_620416.1 | |
NM_138999.1 | 1867 | Intron | NP_620552.1 | |||
XM_005266774.4 | 1867 | Silent Mutation | CTA,TTA | L,L 521 | XP_005266831.1 | |
XM_017026017.1 | 1867 | Silent Mutation | CTA,TTA | L,L 521 | XP_016881506.1 | |
XM_017026018.1 | 1867 | Silent Mutation | CTA,TTA | L,L 521 | XP_016881507.1 | |
XM_017026019.1 | 1867 | Silent Mutation | CTA,TTA | L,L 513 | XP_016881508.1 | |
XM_017026020.1 | 1867 | Silent Mutation | CTA,TTA | L,L 513 | XP_016881509.1 | |
XM_017026021.1 | 1867 | Silent Mutation | CTA,TTA | L,L 379 | XP_016881510.1 | |
XM_017026022.1 | 1867 | Intron | XP_016881511.1 | |||
XM_017026023.1 | 1867 | Intron | XP_016881512.1 | |||
XM_017026024.1 | 1867 | Intron | XP_016881513.1 |