Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGGGAGATCGTGCACATCCAGG[C/T]GGGCCAGTGCGGGAACCAGATCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615103 | ||||||||||||||||||||
Literature Links: |
TUBB6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUBB6 - tubulin beta 6 class V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303524.1 | 260 | Missense Mutation | GCG,GTG | A,V 9 | NP_001290453.1 | |
NM_001303525.1 | 260 | Missense Mutation | GCG,GTG | A,V 9 | NP_001290454.1 | |
NM_001303526.1 | 260 | Missense Mutation | GCG,GTG | A,V 9 | NP_001290455.1 | |
NM_001303527.1 | 260 | UTR 5 | NP_001290456.1 | |||
NM_001303528.1 | 260 | UTR 5 | NP_001290457.1 | |||
NM_001303529.1 | 260 | UTR 5 | NP_001290458.1 | |||
NM_001303530.1 | 260 | UTR 5 | NP_001290459.1 | |||
NM_032525.2 | 260 | Missense Mutation | GCG,GTG | A,V 9 | NP_115914.1 |