Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTTCTTCGAGCTTTTCCAGAAT[A/G]AATTTCCTGAATATGGCACTGTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH19 - cadherin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271028.1 | 2280 | UTR 3 | NP_001257957.1 | |||
NM_021153.3 | 2280 | Silent Mutation | TTC,TTT | F,F 698 | NP_066976.1 | |
XM_011525931.2 | 2280 | Silent Mutation | TTC,TTT | F,F 487 | XP_011524233.1 | |
XM_011525932.1 | 2280 | Intron | XP_011524234.1 | |||
XM_017025711.1 | 2280 | Silent Mutation | TTC,TTT | F,F 487 | XP_016881200.1 |