Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGTGAAGATGGAGATATTCATC[A/G]AGGTTTTCAGTCACTTCTTGTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612086 | ||||||||||||||||||||
Literature Links: |
HMSD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMSD - histocompatibility minor serpin domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123366.1 | 363 | Missense Mutation | CAA,CGA | Q,R 40 | NP_001116838.1 | |
XM_011525930.2 | 363 | Missense Mutation | CAA,CGA | Q,R 40 | XP_011524232.1 | |
XM_017025710.1 | 363 | Missense Mutation | CAA,CGA | Q,R 40 | XP_016881199.1 |