Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGTTTTGGGAAGTGATCAGCGA[C/T]GAGCACGGCATCGACCCGGCCGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBB6 - tubulin beta 6 class V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303524.1 | 312 | Silent Mutation | GAC,GAT | D,D 26 | NP_001290453.1 | |
NM_001303525.1 | 312 | Silent Mutation | GAC,GAT | D,D 26 | NP_001290454.1 | |
NM_001303526.1 | 312 | Silent Mutation | GAC,GAT | D,D 26 | NP_001290455.1 | |
NM_001303527.1 | 312 | Intron | NP_001290456.1 | |||
NM_001303528.1 | 312 | UTR 5 | NP_001290457.1 | |||
NM_001303529.1 | 312 | UTR 5 | NP_001290458.1 | |||
NM_001303530.1 | 312 | UTR 5 | NP_001290459.1 | |||
NM_032525.2 | 312 | Silent Mutation | GAC,GAT | D,D 26 | NP_115914.1 |