Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTGGGTTTTCAGCGTCTAGCC[A/G]TGGGTGATCCAGAGCGACTATCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159430 MIM: 604760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBP - myelin basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025081.1 | 639 | Missense Mutation | ACG,ATG | T,M 194 | NP_001020252.1 | |
NM_001025090.1 | 639 | Missense Mutation | ACG,ATG | T,M 168 | NP_001020261.1 | |
NM_001025092.1 | 639 | Missense Mutation | ACG,ATG | T,M 157 | NP_001020263.1 | |
NM_001025100.1 | 639 | Intron | NP_001020271.1 | |||
NM_001025101.1 | 639 | Missense Mutation | ACG,ATG | T,M 301 | NP_001020272.1 | |
NM_002385.2 | 639 | Missense Mutation | ACG,ATG | T,M 183 | NP_002376.1 | |
XM_011526009.2 | 639 | Intron | XP_011524311.1 | |||
XM_017025778.1 | 639 | Intron | XP_016881267.1 | |||
XM_017025779.1 | 639 | Intron | XP_016881268.1 | |||
XM_017025780.1 | 639 | Intron | XP_016881269.1 |
ZNF236 - zinc finger protein 236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |