Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGCCAGCCCAGGAACTGCTTGA[A/G]CTCGCGCCGCACCGGCCCCGAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf60 - chromosome 19 open reading frame 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRLF1 - cytokine receptor like factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004750.4 | 1367 | Missense Mutation | CTC,TTC | L,F 367 | NP_004741.1 | |
XM_011528422.2 | 1367 | Missense Mutation | CTC,TTC | L,F 345 | XP_011526724.1 | |
XM_011528423.2 | 1367 | Missense Mutation | CTC,TTC | L,F 367 | XP_011526725.1 | |
XM_011528424.2 | 1367 | Missense Mutation | CTC,TTC | L,F 345 | XP_011526726.1 |