Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCTCCGCAGCCCCTCGCCCCC[A/G]AGGGACCAGCCGGCCCAGGAAGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604089 MIM: 604142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCST - hematopoietic cell signal transducer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBID - NFKB inhibitor delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYROBP - TYRO protein tyrosine kinase binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173514.1 | 289 | Missense Mutation | CGG,TGG | R,W 59 | NP_001166985.1 | |
NM_001173515.1 | 289 | Missense Mutation | CGG,TGG | R,W 59 | NP_001166986.1 | |
NM_003332.3 | 289 | Missense Mutation | CGG,TGG | R,W 70 | NP_003323.1 | |
NM_198125.2 | 289 | Missense Mutation | CGG,TGG | R,W 70 | NP_937758.1 |