Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACACTTGGCCAGCGTCGCCTTCA[C/T]CTCTTCTAACAAGTTGCTCAGTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607246 | ||||||||||||||||||||
Literature Links: |
AP3D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP3D1 - adaptor related protein complex 3 delta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261826.1 | 3142 | Missense Mutation | ATG,GTG | M,V 1208 | NP_001248755.1 | |
NM_003938.6 | 3142 | Missense Mutation | ATG,GTG | M,V 1146 | NP_003929.4 | |
XM_006722932.2 | 3142 | Missense Mutation | ATG,GTG | M,V 1196 | XP_006722995.1 | |
XM_017027422.1 | 3142 | Missense Mutation | ATG,GTG | M,V 980 | XP_016882911.1 |
IZUMO4 - IZUMO family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB3A - MOB kinase activator 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |