Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGGTGTTCCTGGGAAGAGGCCA[A/G]GGAGTATTTTGAGGACAACACTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616759 MIM: 616633 MIM: 604429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOSIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOSIP - nitric oxide synthase interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR12 - proline rich 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRG2 - proline rich and Gla domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000951.2 | 565 | Missense Mutation | AAG,AGG | K,R 78 | NP_000942.1 | |
NM_001316335.1 | 565 | Missense Mutation | AAG,AGG | K,R 55 | NP_001303264.1 | |
XM_006723286.2 | 565 | Missense Mutation | AAG,AGG | K,R 78 | XP_006723349.1 | |
XM_011527124.1 | 565 | Missense Mutation | AAG,AGG | K,R 55 | XP_011525426.1 |