Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTTGGTCATCTTTGCAGTCGT[C/T]TTCCTGGTGGGAGTGCTGGGCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 113995 | ||||||||||||||||||||
Literature Links: |
C5AR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5AR1 - complement component 5a receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001736.3 | 163 | Silent Mutation | GTC,GTT | V,V 47 | NP_001727.1 | |
XM_005259190.4 | 163 | UTR 5 | XP_005259247.1 |