Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTTGCTCCAGATGGCCCGGAT[G/T]CCCCCACCATTTCCCCTCTAAACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114890 | ||||||||||||||||||||
Literature Links: |
CEACAM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM5 - carcinoembryonic antigen related cell adhesion molecule 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291484.2 | 831 | Missense Mutation | GCC,TCC | A,S 239 | NP_001278413.1 | |
NM_001308398.1 | 831 | Missense Mutation | GCC,TCC | A,S 239 | NP_001295327.1 | |
NM_004363.5 | 831 | Missense Mutation | GCC,TCC | A,S 239 | NP_004354.3 | |
XM_005258413.3 | 831 | Missense Mutation | GCC,TCC | A,S 239 | XP_005258470.2 | |
XM_011526322.1 | 831 | Intron | XP_011524624.1 | |||
XM_017026145.1 | 831 | Intron | XP_016881634.1 | |||
XM_017026146.1 | 831 | Intron | XP_016881635.1 |