Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACGTTCTCGTAGATGAGGTTTT[C/T]GACATCCTCATACGGGACTTCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602510 MIM: 600782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRH - protein tyrosine phosphatase, receptor type H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161440.2 | 3386 | Missense Mutation | NP_001154912.2 | |||
NM_002842.4 | 3386 | Missense Mutation | NP_002833.4 | |||
XM_011527183.1 | 3386 | Missense Mutation | XP_011525485.1 | |||
XM_011527188.1 | 3386 | Missense Mutation | XP_011525490.1 | |||
XM_011527190.1 | 3386 | Intron | XP_011525492.1 | |||
XM_017027056.1 | 3386 | Missense Mutation | XP_016882545.1 | |||
XM_017027057.1 | 3386 | Missense Mutation | XP_016882546.1 | |||
XM_017027058.1 | 3386 | Missense Mutation | XP_016882547.1 | |||
XM_017027059.1 | 3386 | Missense Mutation | XP_016882548.1 | |||
XM_017027060.1 | 3386 | Missense Mutation | XP_016882549.1 | |||
XM_017027061.1 | 3386 | Missense Mutation | XP_016882550.1 | |||
XM_017027062.1 | 3386 | Missense Mutation | XP_016882551.1 | |||
XM_017027063.1 | 3386 | Missense Mutation | XP_016882552.1 | |||
XM_017027064.1 | 3386 | Intron | XP_016882553.1 |
SYT5 - synaptotagmin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |