Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACAGGGTGCAGCTCTCCAGGGC[A/G]GAGAAGTCAACGGCTAAGAGACCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609235 | ||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032430.1 | 834 | Intron | NP_115806.1 | |||
XM_005259327.3 | 834 | Intron | XP_005259384.1 | |||
XM_011527395.2 | 834 | Intron | XP_011525697.2 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085488.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | NP_001078957.1 | |
NM_001282011.1 | 834 | Silent Mutation | TCC,TCT | S,S 204 | NP_001268940.1 | |
XM_005258812.3 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_005258869.1 | |
XM_011526850.2 | 834 | Intron | XP_011525152.1 | |||
XM_011526851.2 | 834 | Intron | XP_011525153.1 | |||
XM_011526852.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_011525154.1 | |
XM_011526853.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_011525155.1 | |
XM_011526854.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_011525156.1 | |
XM_011526855.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_011525157.1 | |
XM_011526856.2 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_011525158.1 | |
XM_011526857.2 | 834 | Silent Mutation | TCC,TCT | S,S 144 | XP_011525159.1 | |
XM_017026667.1 | 834 | Silent Mutation | TCC,TCT | S,S 204 | XP_016882156.1 | |
XM_017026668.1 | 834 | Silent Mutation | TCC,TCT | S,S 144 | XP_016882157.1 |