Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCACGAGGCTCCTGGCTACCA[A/C]TCGGTGTCCACCAGACTCCAGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPC24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPC24 - SPC24, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317031.1 | 671 | Intron | NP_001303960.1 | |||
NM_001317032.1 | 671 | Intron | NP_001303961.1 | |||
NM_001317033.1 | 671 | Missense Mutation | GAG,GAT | E,D 139 | NP_001303962.1 | |
NM_182513.3 | 671 | Missense Mutation | GAG,GAT | E,D 196 | NP_872319.1 | |
XM_005259753.3 | 671 | Missense Mutation | GAG,GAT | E,D 150 | XP_005259810.1 | |
XM_011527702.1 | 671 | Intron | XP_011526004.1 |