Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCAGGCCCTCCGGGATGGTGT[C/T]CTTCTGTGTCAGCTGCTTAACAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604721 MIM: 164875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2D3A - SH2 domain containing 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAV1 - vav guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258206.1 | 263 | Silent Mutation | GTC,GTT | V,V 41 | NP_001245135.1 | |
NM_001258207.1 | 263 | Silent Mutation | GTC,GTT | V,V 41 | NP_001245136.1 | |
NM_005428.3 | 263 | Silent Mutation | GTC,GTT | V,V 41 | NP_005419.2 | |
XM_005259642.1 | 263 | Silent Mutation | GTC,GTT | V,V 41 | XP_005259699.1 |