Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGGCTTTGATGATGGTCACGGT[A/G]AGGCGCCCGGCCGTGGGGAGGTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600327 | ||||||||||||||||||||
Literature Links: |
SYT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYT3 - synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160328.1 | 1543 | Silent Mutation | CTC,CTT | L,L 448 | NP_001153800.1 | |
NM_001160329.1 | 1543 | Silent Mutation | CTC,CTT | L,L 448 | NP_001153801.1 | |
NM_032298.2 | 1543 | Silent Mutation | CTC,CTT | L,L 448 | NP_115674.1 | |
XM_011527390.2 | 1543 | Silent Mutation | CTC,CTT | L,L 448 | XP_011525692.1 | |
XM_011527391.2 | 1543 | Silent Mutation | CTC,CTT | L,L 448 | XP_011525693.1 |