Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTCCTCTTCTGCTTCCCGGAG[C/T]GCTTGCCGTCGATGTTGAGGCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIRAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIRAS1 - DIRAS family GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145173.3 | 865 | Missense Mutation | CAC,CGC | H,R 181 | NP_660156.1 | |
XM_011527713.2 | 865 | Missense Mutation | CAC,CGC | H,R 215 | XP_011526015.1 |