Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCTGAGCTCCAGCACCATCCA[C/T]TACGAGTGTGTGGATTGCAAGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600283 MIM: 164176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2F2 - POU class 2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF574 - zinc finger protein 574 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022752.5 | 407 | Silent Mutation | CAC,CAT | H,H 125 | NP_073589.4 | |
XM_005259159.2 | 407 | Silent Mutation | CAC,CAT | H,H 125 | XP_005259216.1 | |
XM_011527228.2 | 407 | Silent Mutation | CAC,CAT | H,H 125 | XP_011525530.1 | |
XM_011527229.2 | 407 | Silent Mutation | CAC,CAT | H,H 125 | XP_011525531.1 | |
XM_011527230.2 | 407 | Silent Mutation | CAC,CAT | H,H 125 | XP_011525532.1 | |
XM_017027149.1 | 407 | Silent Mutation | CAC,CAT | H,H 215 | XP_016882638.1 |