Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGCTGCTGCTGCTCCTGGCGC[C/T]GACGTGCCTCCTCATGGGCCCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD4 - bromodomain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014299.2 | 4064 | Intron | NP_055114.1 | |||
NM_058243.2 | 4064 | Missense Mutation | CAG,CGG | Q,R 1281 | NP_490597.1 | |
XM_011527854.1 | 4064 | Missense Mutation | CAG,CGG | Q,R 1281 | XP_011526156.1 | |
XM_011527855.1 | 4064 | Intron | XP_011526157.1 | |||
XM_011527856.2 | 4064 | Intron | XP_011526158.1 |
EPHX3 - epoxide hydrolase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |