Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGTAGGAGAGGCACAGCACGGC[C/T]GAGCTGGCCCTAGGGACACACAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600944 MIM: 609074 MIM: 603002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHPS - deoxyhypusine synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXW9 - F-box and WD repeat domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032301.2 | 980 | Silent Mutation | TCA,TCG | S,S 267 | NP_115677.2 | |
XM_005260096.4 | 980 | Silent Mutation | TCA,TCG | S,S 104 | XP_005260153.1 |
LOC105372280 - uncharacterized LOC105372280 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNPO2 - transportin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |