Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGCCCGCCGGGCCTGCTCCAG[C/G]GCATCCTCGTCCTCTCGGCTCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608749 | ||||||||||||||||||||
Literature Links: |
BRD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRD4 - bromodomain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014299.2 | 4026 | Intron | NP_055114.1 | |||
NM_058243.2 | 4026 | Silent Mutation | GCC,GCG | A,A 1268 | NP_490597.1 | |
XM_011527854.1 | 4026 | Silent Mutation | GCC,GCG | A,A 1268 | XP_011526156.1 | |
XM_011527855.1 | 4026 | Intron | XP_011526157.1 | |||
XM_011527856.2 | 4026 | Intron | XP_011526158.1 |
EPHX3 - epoxide hydrolase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |